Skip to Content
MilliporeSigma
O-Phospho-DL-serine ≥98.0% (NT)

O-Phospho-DL-serine

O-Phospho-DL-serine ≥98.0% (NT)
Synonym(s):
DL-2-Amino-3-hydroxypropanoic acid 3-phosphate, DL-SOP, DL-Serine monophosphoric acid
Empirical Formula (Hill Notation):
C3H8NO6P
CAS Number:
Molecular Weight:
185.07
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Amino Acid
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing