Skip to Content
MilliporeSigma
o-Phenylenediamine analytical standard

o-Phenylenediamine

o-Phenylenediamine analytical standard
Synonym(s):
1,2-Diaminobenzene, 1,2-Phenylenediamine, OPD
Empirical Formula (Hill Notation):
C6H8N2
CAS Number:
Molecular Weight:
108.14

Products

Product Category
Brand
Boiling Point (°C)
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing