Skip to Content
MilliporeSigma
Omeprazole Related Compound B Pharmaceutical Secondary Standard; Certified Reference Material

Omeprazole Related Compound B

Omeprazole Related Compound B Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
5-Methoxy-2-benzimidazolethiol
Empirical Formula (Hill Notation):
C8H8N2OS
CAS Number:
Molecular Weight:
180.23

Products

Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade