Skip to Content
MilliporeSigma
Omeprazole Impurity C Pharmaceutical Secondary Standard; Certified Reference Material

Omeprazole Impurity C

Omeprazole Impurity C Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Omeprazole Sulfide, 5-Methoxy-2-{[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]thio}-1H-benzimidazole, 6-Methoxy-2-{[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]thio}-1H-benzimidazole, Pyrmetazol, Ufiprazole
Empirical Formula (Hill Notation):
C17H19N3O2S
CAS Number:
Molecular Weight:
329.42

Products

Brand
Formula Weight
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1