Skip to Content
MilliporeSigma
Octylamine 99%

Octylamine

Octylamine 99%
Synonym(s):
1-Aminooctane, n-Octylamine, Caprylamine, Octylamine
Linear Formula:
CH3(CH2)7NH2
CAS Number:
Molecular Weight:
129.24

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing