Skip to Content
MilliporeSigma
Octadecylphosphonic acid 97%

Octadecylphosphonic acid

Octadecylphosphonic acid 97%
Synonym(s):
n-Octadecylphosphonic acid, ODPA
Empirical Formula (Hill Notation):
C18H39O3P
CAS Number:
Molecular Weight:
334.47

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing