Skip to Content
MilliporeSigma
O-Benzyl-D-serine ≥99.0%

O-Benzyl-D-serine

O-Benzyl-D-serine ≥99.0%
Synonym(s):
(R)-2-Amino-3-benzyloxypropionic acid
Empirical Formula (Hill Notation):
C10H13NO3
CAS Number:
Molecular Weight:
195.22
EC Number:

Products

Brand
Formula Weight
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing