Skip to Content
MilliporeSigma
O6-Benzylguanine ≥98% (TLC), solid

O6-Benzylguanine

O6-Benzylguanine ≥98% (TLC), solid
Empirical Formula (Hill Notation):
C12H11N5O
CAS Number:
Molecular Weight:
241.25

Products

Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1