
Nylon 6/12

Synonym(s):
Hexamethylenediamine-1,12-dodecanedioic acid polymer, Poly(hexamethylene dodecanediamide)
Linear Formula:
(C12H22O4 · C6H16N2)n
CAS Number:
Products
Brand
Melting Point (°C)
Physical Form
Quality Segment
Polymer Composition
Polymer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing