Skip to Content
MilliporeSigma
N-XantPhos Pd G3 95%

N-XantPhos Pd G3

N-XantPhos Pd G3 95%
Empirical Formula (Hill Notation):
C49H40N2O4P2PdS
CAS Number:
Molecular Weight:
921.29

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing