
N-Vanillylnonanamide

Synonym(s):
Capsaicin synthetic, N-(4-Hydroxy-3-methoxybenzyl)nonanamide, N-Vanillylpelargonamide, Nonanoic acid vanillylamide, Nonylvanylamide, Pelargonic acid vanillylamide, Pseudocapsaicin
Empirical Formula (Hill Notation):
C17H27NO3
CAS Number:
Molecular Weight:
293.40
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Purity
Research Area
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing