Skip to Content
MilliporeSigma
N-(Trimethylsilyl)acetamide 95%

N-(Trimethylsilyl)acetamide

N-(Trimethylsilyl)acetamide 95%
Synonym(s):
TMS acetamide
Linear Formula:
CH3CONHSi(CH3)3
CAS Number:
Molecular Weight:
131.25
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing