Skip to Content
MilliporeSigma
N-trans-caffeoyltyramine ≥85% (LC/MS-ELSD)

N-trans-caffeoyltyramine

N-trans-caffeoyltyramine ≥85% (LC/MS-ELSD)
Synonym(s):
Typheramide
Empirical Formula (Hill Notation):
C17H17NO4
CAS Number:
Molecular Weight:
299.32

Products

Product Category
Brand
Formula Weight
Physical Form
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing