Skip to Content
MilliporeSigma
N-Phenylanthranilic acid 98%

N-Phenylanthranilic acid

N-Phenylanthranilic acid 98%
Synonym(s):
2-(Phenylamino)benzoic acid, Diphenylamine-2-carboxylic acid, DpC, N-Phenylanthranilic acid
Linear Formula:
2-(C6H5NH)C6H4COOH
CAS Number:
Molecular Weight:
213.23

Products

Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Research Area
Quality Segment
Application
Markush Class
Markush Group
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing