Skip to Content
MilliporeSigma
Noroxymorphone hydrochloride solution 1.0 mg/mL in methanol: DMSO (80:20) (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Noroxymorphone hydrochloride solution

Noroxymorphone hydrochloride solution 1.0 mg/mL in methanol: DMSO (80:20) (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C16H17NO4·HCl
CAS Number:
Molecular Weight:
323.77

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique