Skip to Content
MilliporeSigma
Norfloxacin European Pharmacopoeia (EP) Reference Standard

Norfloxacin

Norfloxacin European Pharmacopoeia (EP) Reference Standard
Synonym(s):
1-Ethyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic acid, 1-Ethyl-6-fluoro-1,4-dihydro-4-oxo-7-piperazino-3-quinolinecarboxylic acid, Norfloxacin
Empirical Formula (Hill Notation):
C16H18FN3O3
CAS Number:
Molecular Weight:
319.33

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique