Skip to Content
MilliporeSigma
Nonylamine ≥99.5% (GC)

Nonylamine

Nonylamine ≥99.5% (GC)
Synonym(s):
1-Aminononane, Nonylamine
Linear Formula:
CH3(CH2)8NH2
CAS Number:
Molecular Weight:
143.27

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing