Skip to Content
MilliporeSigma
Nomifensine maleate salt

Nomifensine maleate salt

Nomifensine maleate salt
Synonym(s):
1,2,3,4-Tetrahydro-2-methyl-4-phenyl-8-isoquinolinamine maleate salt
Empirical Formula (Hill Notation):
C16H18N2 · C4H4O4
CAS Number:
Molecular Weight:
354.40
EC Number:

Products

Product Category
Formula Weight
Gene Alias
Physical Form
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing