Skip to Content
MilliporeSigma
N,O-Bis(trimethylsilyl)carbamate ≥98.0% (T)

N,O-Bis(trimethylsilyl)carbamate

N,O-Bis(trimethylsilyl)carbamate ≥98.0% (T)
Synonym(s):
BSC, Trimethylsilyl N-(trimethylsilyl)carbamate
Linear Formula:
(CH3)3SiNHCO2Si(CH3)3
CAS Number:
Molecular Weight:
205.40
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing