Skip to Content
MilliporeSigma
N,O-Bis(trimethylsilyl)acetamide derivatization grade (GC derivatization), LiChropur™, ≥98.5% (GC)

N,O-Bis(trimethylsilyl)acetamide

N,O-Bis(trimethylsilyl)acetamide derivatization grade (GC derivatization), LiChropur™, ≥98.5% (GC)
Synonym(s):
BSA, N,O-Bis(trimethylsilyl)acetamide
Linear Formula:
CH3C[=NSi(CH3)3]OSi(CH3)3
CAS Number:
Molecular Weight:
203.43
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Technique