![N-Nitroso-N-methyl-4-aminobutyric acid analytical standard](/deepweb/assets/sigmaaldrich/product/images/380/880/25ad9a8b-546f-41f7-8fe3-0e7ec23e9b39/640/25ad9a8b-546f-41f7-8fe3-0e7ec23e9b39.jpg)
N-Nitroso-N-methyl-4-aminobutyric acid
![N-Nitroso-N-methyl-4-aminobutyric acid analytical standard](/deepweb/assets/sigmaaldrich/product/images/380/880/25ad9a8b-546f-41f7-8fe3-0e7ec23e9b39/640/25ad9a8b-546f-41f7-8fe3-0e7ec23e9b39.jpg)
Synonym(s):
4-(Methylnitrosoamino)butanoic acid solution, 4-(Methylnitrosoamino)butanoic acid, N-Methyl-N-(3-carboxypropyl)nitrosamine
Empirical Formula (Hill Notation):
C5H10N2O3
CAS Number:
Molecular Weight:
146.14
Products
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing