Skip to Content
MilliporeSigma
N,N-Dimethylmethyleneiminium iodide 98%

N,N-Dimethylmethyleneiminium iodide

N,N-Dimethylmethyleneiminium iodide 98%
Synonym(s):
N,N-Dimethylmethyleneammonium iodide, Eschenmoser’s salt
Linear Formula:
CH2=N+(CH3)2I-
CAS Number:
Molecular Weight:
185.01
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Purity
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing