Skip to Content
MilliporeSigma
N,N-Dimethylformamide diisopropyl acetal 95%

N,N-Dimethylformamide diisopropyl acetal

N,N-Dimethylformamide diisopropyl acetal 95%
Synonym(s):
1,1-Diisopropoxy-N,N-dimethylmethylamine, 1,1-Diisopropoxytrimethylamine
Linear Formula:
(CH3)2NCH[OCH(CH3)2]2
CAS Number:
Molecular Weight:
175.27
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing