N,N-Dimethylacetamide dimethyl acetal
Synonym(s):
1,1-Dimethoxy-N,N-dimethylethylamine
Linear Formula:
CH3C(OCH3)2N(CH3)2
CAS Number:
Molecular Weight:
133.19
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing