Skip to Content
MilliporeSigma
N,N′-Bis(salicylidene)ethylenediaminocobalt(II) hydrate 97%

N,N′-Bis(salicylidene)ethylenediaminocobalt(II) hydrate

N,N′-Bis(salicylidene)ethylenediaminocobalt(II) hydrate 97%
Synonym(s):
N,N′-Disalicylidene-ethylenediamine cobalt(II) salt, Co(SALEN)2, Ethylenebis(salicylimine) cobalt(II) salt, Salcomine
Linear Formula:
[CH2N=CHC6H4-2-(O)]2Co·xH2O
CAS Number:
Molecular Weight:
325.23 (anhydrous basis)
EC Number:

Products

Brand
Formula Weight
Physical Form
Purity
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing