Skip to Content
MilliporeSigma
N,N′-Bis(3-methylphenyl)-N,N′-diphenylbenzidine 99%

N,N′-Bis(3-methylphenyl)-N,N′-diphenylbenzidine

N,N′-Bis(3-methylphenyl)-N,N′-diphenylbenzidine 99%
Synonym(s):
TPD
Linear Formula:
[-C6H4-4-N(C6H4CH3)C6H5]2
CAS Number:
Molecular Weight:
516.67
EC Number:

Products

Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing