Skip to Content
MilliporeSigma
N-Methyl-1-deoxynojirimycin ≥98%

N-Methyl-1-deoxynojirimycin

N-Methyl-1-deoxynojirimycin ≥98%
Synonym(s):
1,5-Dideoxy-1,5-imino-1-methyl-D-sorbitol
Empirical Formula (Hill Notation):
C7H15NO4
CAS Number:
Molecular Weight:
177.20

Products

Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Purity
Activity Spectrum
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing