Skip to Content
MilliporeSigma
[Nle4, D-Phe7]-α-Melanocyte Stimulating Hormone trifluoroacetate salt ≥95% (HPLC)

[Nle4, D-Phe7]-α-Melanocyte Stimulating Hormone trifluoroacetate salt

[Nle4, D-Phe7]-α-Melanocyte Stimulating Hormone trifluoroacetate salt ≥95% (HPLC)
Empirical Formula (Hill Notation):
C78H111N21O19 · xC2HF3O2
CAS Number:
Molecular Weight:
1646.85 (free base basis)

Products

Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Sterility
Showing 1-1 of 1