![Nitrofural impurity B European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/606/553/ffc5552f-a9b9-4b91-8479-940bd92f3c3b/640/ffc5552f-a9b9-4b91-8479-940bd92f3c3b.png)
Nitrofural impurity B
![Nitrofural impurity B European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/606/553/ffc5552f-a9b9-4b91-8479-940bd92f3c3b/640/ffc5552f-a9b9-4b91-8479-940bd92f3c3b.png)
Synonym(s):
5-Nitro-2-furaldehyde diacetate, 5-Nitrofurfurylidene diacetate
Empirical Formula (Hill Notation):
C9H9NO7
CAS Number:
Molecular Weight:
243.17
Products
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing