Skip to Content
MilliporeSigma
Nisoxetine hydrochloride solid, ≥98% (HPLC)

Nisoxetine hydrochloride

Nisoxetine hydrochloride solid, ≥98% (HPLC)
Synonym(s):
(±)-γ-(2-Methoxyphenoxy)-N-methyl-benzenepropanamine hydrochloride, LY-94,939
Empirical Formula (Hill Notation):
C17H21NO2 · HCl
CAS Number:
Molecular Weight:
307.82

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing