Skip to Content
MilliporeSigma
Nifedipine Nitrophenylpyridine Analog certified reference material, pharmaceutical secondary standard

Nifedipine Nitrophenylpyridine Analog

Nifedipine Nitrophenylpyridine Analog certified reference material, pharmaceutical secondary standard
Synonym(s):
Oxidized Nifedipine, 2,6-Dimethyl-4-(2´-nitrophenyl)-3,5-pyridinecarboxylic acid dimethyl ester
Empirical Formula (Hill Notation):
C17H16N2O6
CAS Number:
Molecular Weight:
344.32

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade