Skip to Content
MilliporeSigma
Nicergoline analytical standard, for drug analysis

Nicergoline

Nicergoline analytical standard, for drug analysis
Synonym(s):
5-Bromonicotinic acid 10-methoxy-1,6-dimethylergoline-8-methyl ester
Empirical Formula (Hill Notation):
C24H26BrN3O3
CAS Number:
Molecular Weight:
484.39
EC Number:

Products

Brand
Formula Weight
Manufacturer
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2