Skip to Content
MilliporeSigma
N-Hydroxysulfosuccinimide sodium salt ≥98% (HPLC)

N-Hydroxysulfosuccinimide sodium salt

N-Hydroxysulfosuccinimide sodium salt ≥98% (HPLC)
Synonym(s):
Hydroxy-2,5-dioxopyrrolidine-3-sulfonicacid sodium salt, Sulfo-NHS
Empirical Formula (Hill Notation):
C4H4NNaO6S
CAS Number:
Molecular Weight:
217.13

Products

Product Category
Formula Weight
Physical Form
Purity
Chemical Composition
Functional Group
Application
Markush Class
Markush Group
Reaction Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing