Skip to Content
MilliporeSigma
N-Hydroxy-5-norbornene-2,3-dicarboxylic acid imide 97%

N-Hydroxy-5-norbornene-2,3-dicarboxylic acid imide

N-Hydroxy-5-norbornene-2,3-dicarboxylic acid imide 97%
Synonym(s):
N-Hydroxybicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid imide, HONB
Empirical Formula (Hill Notation):
C9H9NO3
CAS Number:
Molecular Weight:
179.17
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing