Skip to Content
MilliporeSigma
Neurotensin Fragment 8-13 acetate salt ≥97% (HPLC)

Neurotensin Fragment 8-13 acetate salt

Neurotensin Fragment 8-13 acetate salt ≥97% (HPLC)
Empirical Formula (Hill Notation):
C38H64N12O8
CAS Number:
Molecular Weight:
816.99

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Application
Technique
Showing 1-1 of 1