Skip to Content
MilliporeSigma
N-Ethyldiisopropylamine solution suitable for peptide synthesis, ~2 M in 1-methyl-2-pyrrolidinone

N-Ethyldiisopropylamine solution

N-Ethyldiisopropylamine solution suitable for peptide synthesis, ~2 M in 1-methyl-2-pyrrolidinone
Synonym(s):
N,N-Diisopropylethylamine, DIPEA, Hünig’s base
Empirical Formula (Hill Notation):
C8H19N
CAS Number:
Molecular Weight:
129.24

Products

Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1