Neotame
Synonym(s):
N-[N-(3,3-Dimethylbutyl)-L-α-aspartyl]-L-phenylalanine 1-methyl ester
Empirical Formula (Hill Notation):
C20H30N2O5
CAS Number:
Molecular Weight:
378.46
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing