Nebivolol hydrochloride
Synonym(s):
Narbivolol, Nebilet, R-65824, rel-(aR,a′R,2R,2′S)-a,a′-[iminobis(methylene)]bis[6-fluoro-3,4-dihydro-2H-1-benzopyran-2-methanol] hydrochloride, Nebivolol hydrochloride
Empirical Formula (Hill Notation):
C22H25F2NO4 · HCl
CAS Number:
Molecular Weight:
441.90
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3