Skip to Content
MilliporeSigma
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate

N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate

N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate
Synonym(s):
3-(N,N-Dimethyldodecylammonio)propanesulfonate, 3-(N,N-Dimethyllaurylammonio)propanesulfonate, 3-(Dodecyldimethylammonio)propanesulfonate, 3-(Lauryldimethylammonio)propanesulfonate, Dodecyldimethyl(3-sulfopropyl)ammonium hydroxide inner salt, Lauryl sulfobetaine, SB3-12
Linear Formula:
CH3(CH2)11N+(CH3)2CH2CH2CH2SO3-
CAS Number:
Molecular Weight:
335.55
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Application