Skip to Content
MilliporeSigma
N-(Diphenylmethylene)glycine ethyl ester 98%

N-(Diphenylmethylene)glycine ethyl ester

N-(Diphenylmethylene)glycine ethyl ester 98%
Synonym(s):
Ethyl (diphenylmethylenamino)acetate
Linear Formula:
(C6H5)2C=NCH2CO2C2H5
CAS Number:
Molecular Weight:
267.32

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing