![N-(+)-Biotinyl-6-aminohexanoic acid ≥97.0% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/141/434/212340da-0665-4308-a374-e9f25755d0ff/640/212340da-0665-4308-a374-e9f25755d0ff.png)
N-(+)-Biotinyl-6-aminohexanoic acid
![N-(+)-Biotinyl-6-aminohexanoic acid ≥97.0% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/141/434/212340da-0665-4308-a374-e9f25755d0ff/640/212340da-0665-4308-a374-e9f25755d0ff.png)
Synonym(s):
(+)-Biotin-ε-aminocaproic acid, 6-((Biotinoyl)amino)hexanoic acid, Biotin-X, N-(+)-Biotinyl-6-aminocaproic acid
Empirical Formula (Hill Notation):
C16H27N3O4S
CAS Number:
Molecular Weight:
357.47
Products
Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing