Skip to Content
MilliporeSigma
N-Cyclohexyl-N′-(2-morpholinoethyl)carbodiimide methyl-p-toluenesulfonate 95%

N-Cyclohexyl-N′-(2-morpholinoethyl)carbodiimide methyl-p-toluenesulfonate

N-Cyclohexyl-N′-(2-morpholinoethyl)carbodiimide methyl-p-toluenesulfonate 95%
Synonym(s):
4-[2-[(Cyclohexylcarbonimidoyl)amino]ethyl]-4-methylmorpholinium p-toluenesulfonate, N-Cyclohexyl-N′-(β-[N-methylmorpholino]ethyl)carbodiimide p-toluenesulfonate salt, CMC, CMC metho-p-toluenesulfonate, Morpho CDI
Empirical Formula (Hill Notation):
C14H26N3O · C7H7O3S
CAS Number:
Molecular Weight:
423.57
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Showing 1-1 of 1