Skip to Content
MilliporeSigma
vibrant-m

N-[(cis-4-Isopropylcyclohexyl)carbonyl]-D-phenylalanine

vibrant-m
Synonym(s):
N-[[cis-4-(1-Methylethyl)cyclohexyl]carbonyl]-D-phenylalanine
Empirical Formula (Hill Notation):
C19H27NO3
CAS Number:
Molecular Weight:
317.42

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade