Skip to Content
MilliporeSigma
N-Boc-L-prolinol 98%

N-Boc-L-prolinol

N-Boc-L-prolinol 98%
Synonym(s):
(S)-1-Boc-2-pyrrolidinemethanol
Empirical Formula (Hill Notation):
C10H19NO3
CAS Number:
Molecular Weight:
201.26

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing