Skip to Content
MilliporeSigma
N-Boc-ethylenediamine ≥98.0% (NT)

N-Boc-ethylenediamine

N-Boc-ethylenediamine ≥98.0% (NT)
Synonym(s):
N-Boc-1,2-diaminoethane, tert-Butyl N-(2-aminoethyl)carbamate
Linear Formula:
(CH3)3COCONHCH2CH2NH2
CAS Number:
Molecular Weight:
160.21

Products

Product Category
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing