Skip to Content
MilliporeSigma
N-Boc-cis-4-hydroxy-L-proline methyl ester 97%

N-Boc-cis-4-hydroxy-L-proline methyl ester

N-Boc-cis-4-hydroxy-L-proline methyl ester 97%
Empirical Formula (Hill Notation):
C11H19NO5
CAS Number:
Molecular Weight:
245.27

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing