Skip to Content
MilliporeSigma
N-Boc-1,6-hexanediamine ≥98.0% (NT)

N-Boc-1,6-hexanediamine

N-Boc-1,6-hexanediamine ≥98.0% (NT)
Synonym(s):
N-Boc-1,6-diaminohexane, tert-Butyl-N-(6-aminohexyl)carbamate
Empirical Formula (Hill Notation):
C11H24N2O2
CAS Number:
Molecular Weight:
216.32

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Reagent Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing