Skip to Content
MilliporeSigma
N-Benzylhydroxylamine hydrochloride puriss., ≥99.0% (AT)

N-Benzylhydroxylamine hydrochloride

N-Benzylhydroxylamine hydrochloride puriss., ≥99.0% (AT)
Linear Formula:
C6H5CH2NHOH · HCl
CAS Number:
Molecular Weight:
159.61

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing