Skip to Content
MilliporeSigma
N-Benzylcinchoninium chloride 98%

N-Benzylcinchoninium chloride

N-Benzylcinchoninium chloride 98%
Synonym(s):
9-Hydroxy-1-(phenylmethyl)cinchonanium chloride, BCNC
Empirical Formula (Hill Notation):
C26H29ClN2O
CAS Number:
Molecular Weight:
420.97
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing