Skip to Content
MilliporeSigma
N-(Benzoyloxy)succinimide 96%

N-(Benzoyloxy)succinimide

N-(Benzoyloxy)succinimide 96%
Synonym(s):
Benzoic acid N-hydroxysuccinimide ester
Empirical Formula (Hill Notation):
C11H9NO4
CAS Number:
Molecular Weight:
219.19

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing